1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Ethyl-3-methylimidazolium dicyanamide, 98%, Thermo Scientific Chemicals
CAS: 370865-89-7 | C8H11N5 | 177.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Catalog number ALFH26901.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS110-60-1
IUPAC Namebutane-1,4-diamine
Molecular FormulaC4H12N2
InChI KeyKIDHWZJUCRJVML-UHFFFAOYSA-N
SMILESNCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4550 to 1.4590 (20°C, 589 nm)
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.
Solubility
Soluble in water. Soluble in solvents like methanol, acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Joan Fuller,; Richard T. Carlin,; Hugh C. De Long,; Dustin Haworth. Structure of 1-ethyl-3-methylimidazolium hexafluorophosphate: model for room temperature molten salts. J. Chem. Soc., Chem. Commun.1994 , (3),299-300.
  2. authors. title . Journal name. year of publication , volume issue , pg no.