2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichloro-5-methoxyaniline, 98%, Thermo Scientific Chemicals
CAS: 98446-49-2 | C7H7Cl2NO | 192.039 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALFB25497.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1668-10-6
IUPAC Namehydrogen 2-aminoacetamide chloride
Molecular FormulaC2H7ClN2O
InChI KeyWKNMKGVLOWGGOU-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Clarity0.5% w/v solution in water is transparent to almost transparent.
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%
Assay (Titration ex Chloride)≥97.5 to ≤102.5%