Sodium phenoxide, 98%, Thermo Scientific Chemicals
Sodium phenoxide, 98%, Thermo Scientific Chemicals
Sodium phenoxide, 98%, Thermo Scientific Chemicals
Sodium phenoxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium phenoxide, 98%, Thermo Scientific Chemicals
CAS: 139-02-6 | C6H5NaO | 116.10 g/mol
Have Questions?
Catalog NumberQuantity
ALFB25002.1850 g
Catalog number ALFB25002.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7782-99-2
IUPAC Namesulfurous acid
Molecular FormulaH2O3S
InChI KeyLSNNMFCWUKXFEE-UHFFFAOYSA-N
SMILESOS(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Residue on ignition0.005%
Impurity contentCl 5ppm
Trace Metals(as Pb) 2ppm
Assay (unspecified)6.0% SO₂ min
Impurity contentFe 5ppm
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.
Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.
Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Branchini, B. R.; Behney, C. E.; Southworth, T. L.; Fontaine, D. M.; Gulick, A. M.; Vinyard, D. J.; Brudvig, G. W. Experimental Support for a Single Electron-Transfer Oxidation Mechanism in Firefly Bioluminescence. J. Am. Chem. Soc. 2015, 137 (24), 7592-7595.
  2. Zhang, W.; Fan, X.; Tian, W.; Chen, H.; Zhu, X.; Zhang, H. Preparation of a P(THF-co-PO)-b-PB-b-P(THF-co-PO) triblock copolymer via cationic ring-opening polymerization and its use as a thermoset polymer. RSC Adv. 2015, 5 (81), 66073-66081.