3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Fluorophenyl isothiocyanate, 97+%, Thermo Scientific Chemicals
CAS: 404-72-8 | C7H4FNS | 153.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFB25001.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Fluorophenyl isothiocyanate
CAS404-72-8
Health Hazard 1H227-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c