3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromobenzylamine, 95%, Thermo Scientific Chemicals
CAS: 10269-01-9 | C7H8BrN | 186.052 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24963.065 g
Catalog number ALFB24963.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS122-88-3
IUPAC Name2-(4-chlorophenoxy)acetic acid
Molecular FormulaC8H7ClO3
InChI KeySODPIMGUZLOIPE-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Melting point>=156°C
Water=<1 %
Titration with NaOH>=97.5 %
Appearance (Form)Powder
View more
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromobenzylamine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agro chemicals, dyestuff.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
RUO – Research Use Only
General References:
  1. Jeremy P. Mallari; Anang A. Shelat; Aaron Kosinsk; Conor R. Caffrey; Michele Connelly; Fangyi Zhu; James H. McKerrow and R. Kiplin Guy. Structure-Guided Development of Selective TbcatB Inhibitors. J. Med. Chem. 2009, 52 (20), 6489-6493.
  2. Aldo Caiazzo; Paula M. L. Garcia; Ron Wever; Jan C. M. van Hest; Alan E. Rowan and Joost N. H. Reek. Synergy between chemo- and bio-catalysts in multi-step transformations. Organic & Biomolecular Chemistry. 2009, 14 (7), 2926-2932.