p-Tolylacetylene, 98%, Thermo Scientific Chemicals
p-Tolylacetylene, 98%, Thermo Scientific Chemicals
p-Tolylacetylene, 98%, Thermo Scientific Chemicals
p-Tolylacetylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Tolylacetylene, 98%, Thermo Scientific Chemicals
CAS: 766-97-2 | C9H8 | 116.163 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24812.065 g
Catalog number ALFB24812.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Tolylacetylene
CAS766-97-2
Health Hazard 1H226-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H304-H226-H319-H335
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P374-P380-P501c
View more
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.
Solubility
Insoluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
RUO – Research Use Only
General References:
  1. Yanxia Zhao; Yanyan Liu; Yibo Lei; Biao Wu; Xiao-Juan Yang. Activation of alkynes by an α-diimine-stabilized Al-Al-bonded compound: insertion into the Al-Al bond or cycloaddition to AlN2C2 rings. Chemical Communications. 2013, 49 (40), 4546-4548.
  2. Amit N. Pandya; James T. Fletcher; Eric M. Villa; Devendra K. Agrawal. Silver-Mediated Synthesis of Indolizines via Oxidative C-H functionalization and 5- endo-dig cyclization. Tetrahedron Letters. 2014, 55 (50), 6922-6924.