5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%, Thermo Scientific Chemicals
CAS: 1046-56-6 | C20H14N4 | 310.36 g/mol
Have Questions?
Catalog NumberQuantity
ALFB24601.031 g
Catalog number ALFB24601.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS768-66-1
IUPAC Name2,2,6,6-tetramethylpiperidine
Molecular FormulaC9H19N
InChI KeyRKMGAJGJIURJSJ-UHFFFAOYSA-N
SMILESCC1(C)CCCC(C)(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to light brown
Refractive index1.4425 to 1.4465 (20°C, 589 nm)
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine is widely utilized as a reagent for the determination of sub microgram levels of iron in serum. It is also used for the simultaneous determination of iron and copper.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Reagent for the determination of Cu, and for the spectrophotometric determination of Fe(II) and Ru(II): Anal. Chem., 46, 992 (1974); Anal. Chim. Acta, 78, 423 (1975).
  2. Yamada, M.; Muto, Y.; Kurihara, H.; Slanina, Z.; Suzuki, M.; Maeda, Y.; Rubin, Y.; Olmstead, M. M.; Balch, A. L.; Nagase, S.; Lu, X.; Akasaka, T. Regioselective Cage Opening of La2@D2(10611)-C72 with 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine. Angew. Chem. Int. Ed. 2015, 54 (7), 2232-2235.
  3. Anjomshoa, M.; Hadadzadeh, H.; Mahani, M. T.; Fatemi, S. J.; Sardou, M. A.; Rudbari, H. A.; Nardo, V. M. A mononuclear Cu(II) complex with 5,6-diphenyl-3-(2-pyridyl)-1,2,4-triazine: Synthesis, crystal structure, DNA- and BSA-binding, molecular modeling, and anticancer activity against MCF-7, A-549, and HT-29 cell lines. Angew. Chem. Int. Ed. 2015, 96, 66-82.