Furfurylamine, 99%, Thermo Scientific Chemicals
Furfurylamine, 99%, Thermo Scientific Chemicals
Furfurylamine, 99%, Thermo Scientific Chemicals
Furfurylamine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Furfurylamine, 99%, Thermo Scientific Chemicals
CAS: 617-89-0 | C5H7NO | 97.117 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23975.22100 g
Catalog number ALFB23975.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS90-33-5
IUPAC Name7-hydroxy-4-methyl-2H-chromen-2-one
Molecular FormulaC10H8O3
InChI KeyHSHNITRMYYLLCV-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC2=CC(O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH96.0 to 104.0 % (On dry substance)
Water=<2.5 % (K.F.)
Appearance (Color)White to light yellow to light beige
Melting point186°C to 191°C
HPLC>=96.0 %
View more
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, air, acids.
RUO – Research Use Only
General References:
  1. Kuo Yueh-Hsiung;Shih Kae-Shyang. A new method for preparation of 3-hydroxypyridines from furfurylamines by photooxygenation. Pharmaceutical Bulletin. 1991, 39 (1), 181-183.
  2. Rajappa Murali; H Surya Prakash Rao; Hans W Scheeren. Intra-molecular Diels-Alder reactions of citraconamic acids from furfurylamines and citraconic anhydride: effects of substitution in the furan ring on regioselectivity. Tetrahedron. 2001, 57 (15), 3165-3174.