1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methylpyrrolidine, 98%, Thermo Scientific Chemicals
CAS: 120-94-5 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFB23799.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS79-06-1
IUPAC Nameprop-2-enamide
Molecular FormulaC3H5NO
InChI KeyHRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILESNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤2.0%
Melting Point (clear melt)79-87?C
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Appearance (Color)White
View more
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Methylpyrrolidine is a methylated pyrollidine and is involved as an essential part of the structure of cefipime. It is also an active component of cigarette smoke.
Solubility
Miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
  1. Pohlmann, S.; Olyschlager, T.; Goodrich, P.; Vicente, J. A.; Jacquemin, J.; Balducci, A. Azepanium-based ionic liquids as green electrolytes for high voltage supercapacitors. J. Power Sources 2015, 273, 931-936.
  2. Dabrowska, M.; Starek, M.; Krzek, J.; Papp, E.; Krol, P. A degradation study of cefepime hydrochloride in solutions under various stress conditions by TLC-densitometry. Biomed. Chromatogr. 2015, 29 (3), 388-395.