6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals
6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals
6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals
6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
6-Hydroxyflavanone, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFB23755.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS37529-27-4
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow to yellow to orange
Assay (GC)≥97.5%
Refractive Index1.5185-1.5225 @ 20?C