2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
CAS: 25013-16-5 | C11H16O2 | 180.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALFB23530.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS9005-38-3
Molecular Formula(C6H7O7)A(C6H7O7)BNa
SMILES[Na+].O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C([O-])=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C([O-])=O
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Assay (special)Lead: ≤5 ppm
Loss on Drying≤15.0%
Viscosity1000 to 1500 mPa's (1% solution)
FormPowder
Assay (special)Arsenic: ≤3 ppm
View more
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.
Notes
Store in the dark place. Store away from light, oxidizing agents.
RUO – Research Use Only
General References:
  1. JM. Alauddin; Ronald E. Verrall. Apparent molal volume studies of 2,6-di-tert-butyl-4-methylphenol, 2-tert-butyl-4-methoxyphenol, and 2,6-di-tert-butyl-4-(hydroxymethyl)phenol in aqueous micelle solutions of sodium dodecanoate as a function of micelle concentration and temperature. J. Phys. Chem. 1984, 88, (23), 5725-5730.
  2. Yasuo Butsugan; Masao Muto; Masao Kawai; Shuki Araki; Yoshiaki Murase; Kazuki Saito. Structures of autoxidation products of 2-tert-butyl-4-methoxyphenol in aqueous alkaline solution. J. Org. Chem. 1989, 54 (17), 4215-4217.