n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Dodecylpyridinium chloride hydrate, 98%, Thermo Scientific Chemicals
CAS: 207234-02-4 | C17H30ClN | 283.88 g/mol
Have Questions?
Catalog NumberQuantity
ALFB23492.22100 g
Catalog number ALFB23492.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS53344-74-4
IUPAC Name2,2'-dichloro-4,4'-bipyridine
Molecular FormulaC10H6Cl2N2
InChI KeyKSELKNWXBOXHEV-UHFFFAOYSA-N
SMILESClC1=NC=CC(=C1)C1=CC(Cl)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Appearance (Color)Cream to pale yellow
Melting Point (clear melt)238.0-250.0?C