Thiotriazinone, 98+%, Thermo Scientific Chemicals
Thiotriazinone, 98+%, Thermo Scientific Chemicals
Thiotriazinone, 98+%, Thermo Scientific Chemicals
Thiotriazinone, 98+%, Thermo Scientific Chemicals
Thiotriazinone, 98+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB22663.0910 g
Catalog number ALFB22663.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1338-43-8
IUPAC Name2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate
Molecular FormulaC24H44O6
InChI KeyNWGKJDSIEKMTRX-NZAQQJATSA-N
SMILESCCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofAFatty acid content: ≥70%
Acid Value5.0-6.0
FormLiquid or viscous liquid
Assay from Suppliers CofASaponification value: ≥150
Appearance (Color)Yellow to very dark yellow and brown-yellow to very dark brown