Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Perfluorodecanoic acid, 97%, Thermo Scientific Chemicals
CAS: 335-76-2 | C10HF19O2 | 514.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFB22292.065 g
Catalog number ALFB22292.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2244-16-8
IUPAC Name(5S)-2-methyl-5-(prop-1-en-2-yl)cyclohex-2-en-1-one
Molecular FormulaC10H14O
InChI KeyULDHMXUKGWMISQ-VIFPVBQESA-N
SMILESCC(=C)[C@H]1CC=C(C)C(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
Optical Rotation58.0 ± 2.0? (neat)
Refractive Index1.4955-1.5030 @ 20?C
Appearance (Color)Clear colorless to white to yellow