alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals
alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals
alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals
alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Toluenesulfonyl fluoride, 99%, Thermo Scientific Chemicals
CAS: 329-98-6 | C7H7FO2S | 174.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALFB22146.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS81290-20-2
IUPAC Nametert-butyl(trifluoromethyl)silane
Molecular FormulaC5H11F3Si
InChI KeyMTYSDPUZDZURPP-UHFFFAOYSA-N
SMILESCC(C)(C)[SiH2]C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
Assay by NMR≥94.0% (1H-NMR, CDCl3)
Water Content (Karl Fischer Titration)≤2.50%
View more
α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
α-Toluenesulfonyl fluoride (PMSF) is used in the preparation of cell lysates to inhibit protein digestion. It is a widely used fatty acid amid hydrolase (FAAH) inhibitor for pretreatment of brain membrane preparations when testing CB1 receptor activity.
Solubility
Soluble in ether, benzene, anhydrous ethanol, methanol, 2- propane. Hydrolyses in water.
Notes
Moisture sensitive. Store away from strong oxidizing agents, amines, bases, water/ moisture. Store at 4°C
RUO – Research Use Only
General References:
  1. C Pinsky; A K Dua; F S LaBella. Phenylmethylsulfonyl fluoride (PMSF) given systemically produces naloxone-reversible analgesia and potentiates effects of beta-endorphin given centrally. Life Sciences. 1982, 31 (12-13), 1193-1196.
  2. G T James.Inactivation of the protease inhibitor phenylmethylsulfonyl fluoride in buffers. Analytical Biochemistry. 1978, 86 (2), 574-579.