1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals
1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals
1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals
1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Methyl-2-phenylindole, 99%, Thermo Scientific Chemicals
CAS: 3558-24-5 | C15H13N | 207.276 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALFB22105.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS367-22-6
IUPAC Name4-chloro-3-fluoroaniline
Molecular FormulaC6H5ClFN
InChI KeyACMJJQYSPUPMPN-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or grey
Melting Point (clear melt)58.0-62.0?C
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder or lumpy/fused solid