4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Diiodobiphenyl, 99%, Thermo Scientific Chemicals
CAS: 3001-15-8 | C12H8I2 | 406.005 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFB21970.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS532-32-1
IUPAC Namesodium benzoate
Molecular FormulaC7H5NaO2
InChI KeyWXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=99.0 % (On dried substance)
Acidity/alkalinity=<0.2 ml 0.1 M NaOH or HCl per gram
Appearance (Form)Crystalline powder or crystals or granules or
Infrared spectrumConforms
Water=<2.0 % (K.F.)
View more
4,4?-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4?-diiodobipheny enhanced the expression of the luciferase gene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4,4′-diiodobiphenyl was coupled with (trimethylsilyl) acetylene via a Sonogashira-Hagihara reaction to afford the (trimethylsilyl)ethynyl derivative. 4,4′-diiodobipheny enhanced the expression of the luciferase gene.
Solubility
Soluble in hot Toluene(very faint turbidity).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only
General References:
  1. Toshiko Yamada-Okabe,; Haruya Sakai,; Yuji Kashima,; Hisafumi Yamada-Okab. Modulation at a cellular level of the thyroid hormone receptor-mediated gene expression by 1,2,5,6,9,10-hexabromocyclododecane (HBCD), 4,4'-diiodobiphenyl (DIB), and nitrofen (NIP). Toxicology Letters. 2005, 155 (1), 127-133.
  2. D. Britton . 4,4'-Diiodobiphenyl. Acta Cryst. 2005, E61, o187-o188.