1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals
1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Iodo-3-nitrobenzene, 99%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALFB21605.1425 g
Catalog number ALFB21605.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS105-45-3
IUPAC Namemethyl 3-oxobutanoate
Molecular FormulaC5H8O3
InChI KeyWRQNANDWMGAFTP-UHFFFAOYSA-N
SMILESCOC(=O)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (Silylated GC)≥98.5%
Refractive Index1.4175-1.4215 @ 20?C