Glybenzcyclamide, 99%, Thermo Scientific Chemicals
Glybenzcyclamide, 99%, Thermo Scientific Chemicals
Glybenzcyclamide, 99%, Thermo Scientific Chemicals
Glybenzcyclamide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glybenzcyclamide, 99%, Thermo Scientific Chemicals
CAS: 10238-21-8 | C23H28ClN3O5S | 494.003 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21459.22100 g
Catalog number ALFB21459.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6334-18-5
IUPAC Name2,3-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyLLMLNAVBOAMOEE-UHFFFAOYSA-N
SMILESClC1=CC=CC(C=O)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point60°C to 64°C
Appearance (Form)Powder or crystals and/or chunks
GC>=98.5 %
Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 M).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glyburide (Glibenclamide) is a sulfonylurea compound that modulates insulin production. Sulfonylureas bind to ATP-dependent K+ channels in beta cells of the pancreas, depolarizing them and stimulating the release of Ca2+, which in turn stimulates insulin production. Glyburide (Glibenclamide) is an ATP-dependent K+ channel (KIR6, KATP) and CFTR Cl- channel blocker. This compound inhibits recombinant CFTR Cl- channels (IC50 = 20 μM).
Solubility
Soluble in ethanol (5 mg/mL), DMSO (25 mg/mL), chloroform (1:36), methanol (1:250), and DMF. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. DG Dills.; J Schneider. Clinical evaluation of glimepiride versus glyburide in NIDDM in a double-blind comparative study. Glimepiride/Glyburide Research Group. Hormone and metabolic research= Hormone. 199628 (9), 426-429.
  2. M Jabůrek.; V Yarov-Yarovoy.; P Paucek. State-dependent inhibition of the mitochondrial KATP channel by glyburide and 5-hydroxydecanoate. Hormone and metabolic research= Hormone. 1998273 (9), 13578-13582.