Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
CAS: 74568-07-3 | C28H24O4 | 424.496 g/mol
Have Questions?
Catalog NumberQuantity
ALFB21320.065 g
Catalog number ALFB21320.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCalix[4]arene
CAS74568-07-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Functionalized calix[4]arenes can bind DNA and induce cell transfection. Calixarenes lend themselves well to many applications because of the multiplicity of options for such structural elaboration such as in electrochemical sensors, optical sensors, chiral recognition devices,
Solubility
Soluble in chloroform.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Kenny B. Lipkowitz and Greg Pearl. Structural features of solid-state calix[4]arene in the cone conformation.J. Org. Chem.,199358(24), 6729-6736.
  2. Sergio Sanz, et al. Calix[4]arene-supported rare earth octahedral.Chem. Commun., 201248(10), 1449-1451.