3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals
3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals
3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals
3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4,5-Trihydroxybenzamide, 98%, Thermo Scientific Chemicals
CAS: 618-73-5 | C7H7NO4 | 169.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFB20620.06
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material3,4,5-Trihydroxybenzamide
CAS618-73-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c