o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals
o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals
o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals
o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals
CAS: 88-19-7 | C7H9NO2S | 171.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number ALFB20544.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS12064-62-9
IUPAC Namedigadolinium(3+) trioxidandiide
Molecular FormulaGd2O3
InChI KeyCMIHHWBVHJVIGI-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Gd+3].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Total conc. of imp.as REO=<0.1 %
Appearance (Form)Powder
o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.
Solubility
Soluble in water (1.6 g/L at 25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases and acids.
RUO – Research Use Only
General References:
  1. JL Kacprzak. Determination of o-toluenesulfonamide in artificial sweeteners containing saccharin. Journal-Association of Official Analytical Chemists. 197861 (6), 1528-1532.
  2. L Xu.; H Shu.; Y Liu.; S Zhang.; ML Trudell. Oxidative cyclization of N-alkyl-o-methyl-arenesulfonamides to biologically important saccharin derivatives. Tetrahedron. 200661 (6), 1528-1532.