5-Chlorothiophene-2-sulfonamide, 97%, Thermo Scientific Chemicals
5-Chlorothiophene-2-sulfonamide, 97%, Thermo Scientific Chemicals
5-Chlorothiophene-2-sulfonamide, 97%, Thermo Scientific Chemicals
5-Chlorothiophene-2-sulfonamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Chlorothiophene-2-sulfonamide, 97%, Thermo Scientific Chemicals
CAS: 53595-66-7 | C4H4ClNO2S2 | 197.65 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20477.031 g
Catalog number ALFB20477.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS865-49-6
IUPAC Nametrichloro(²H)methane
Molecular FormulaCHCl3
InChI KeyHEDRZPFGACZZDS-MICDWDOJSA-N
SMILES[2H]C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
D-Enrichment>=99.95 %
Appearance (Color)Clear colorless
Water=<0.01 % (Coulometric)