4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals
4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Dimethylaminobenzophenone, 98%, Thermo Scientific Chemicals
CAS: 530-44-9 | C15H15NO | 225.291 g/mol
Have Questions?
Catalog NumberQuantity
ALFB20389.065 g
Catalog number ALFB20389.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Dimethylaminobenzophenone
CAS530-44-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-(Dimethylamino)benzophenone is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
General References:
  1. Eliane Chesneau; D.C. Neckers. Electron transfer sensitized photobleaching of rose bengal induced by triplet benzophenones. Journal of Photochemistry and Photobiology A: Chemistry. 1988, 42 (2-3), 269-281.
  2. Edwin M. Kaiser; Jimmie D. Petty. Preparation and condensations of 3-lithiomethylquinoline. J. Org. Chem. 1976, 41 (4), 716-717.