Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium tert-butoxide, 97%, Thermo Scientific Chemicals
CAS: 865-48-5 | C4H9NaO | 96.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALFA19942.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium tert-butoxide
CAS865-48-5
Health Hazard 1H228-H314-H500-X
Health Hazard 2GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Health Hazard 3P210-P235+P410-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
RUO – Research Use Only
General References:
  1. Preferred base, superior to KO-t-Bu or LiO-t-Bu, for promoting Buchwald's Pd catalyzed conversion of aryl bromides to arylamines: Angew. Chem. Int. Ed., 34, 1316, 1348 (1995); J. Org. Chem., 61, 7240 (1996); Tetrahedron Lett., 39, 2219 (1998).
  2. Zhang, M. X.; Hu, X. H.; Xu, Y. H.; Loh, T. P. Selective Dealkylation of Alkyl Aryl Ethers. Asian J. Org. Chem. 2015, 4 (10), 1047-1049.
  3. Kumar, C. V.; Raptis, D.; Koukaras, E. N.; Sygellou, L.; Lianos, P. Study of an indoline-phenothiazine based organic dye for Dye-Sensitized Solar Cells. Theoretical calculations and experimental data. Org. Electron. 2015, 25, 66-73.