Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals
CAS: 67-03-8 | C12H18Cl2N4OS | 337.263 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Catalog number ALFA19560.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThiamine hydrochloride
CAS67-03-8
Health Hazard 1H319-H500
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280i-P305+P351+P338
View more
Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.
Solubility
Soluble in water, glycerol, ethanol, dimethyl sulfoxide and methanol. Insoluble in benzene, hexane, chloroform and ether.
Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Yan, X.; Li, S.; Pan, Y. X.; Yang, Z.; Liu, X. A simple approach to the synthesis of Cu1. 8S dendrites with thiamine hydrochloride as a sulfur source and structure-directing agent. Beilstein J. Nanotechnol. 2015, 6 (1), 881-885.
  2. Sarker, S.; Tammi, R. S.; Biswas, S.; Zaman, S.; Seraj, Z. I. Cloning and Characterization of a Tissue Specific Promoter GluB-1 from Nipponbare by Transformation in Rice(Vitamin-B1) Application. Biores Comm. 2015, 1 (2), 69-75.