Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Styrene, 99%, stab. with 10-15ppm 4-tert-butylcatechol, Thermo Scientific Chemicals
CAS: 100-42-5 | C8H8 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 mL
500 mL
Catalog number ALFA18481.0F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialStyrene
Name Notestabilized with 10 to 15ppm 4-tert-butylcatechol
CAS100-42-5
Health Hazard 1H226-H304-H315-H319-H332-H335-H336-H351-H361d-H372
Health Hazard 2GHS H Statement
H372-H304-H361-H226-H332-H315-H319-H335
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Styrene is used in the preparation of polystyrene which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include refrigerator liners, medical devices and small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN), which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.
Solubility
Miscible with carbon disulfide, ether, methanol, benzene, toluene, ethanol, n-heptane, carbon tetrachloride and acetone. Immiscible with water.
Notes
Air and light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and copper.
RUO – Research Use Only
General References:
  1. Fu, G. D.; Shang, Z.; Hong, L.; Kang, E. T.; Neoh, K. G. Preparation of cross-linked polystyrene hollow nanospheres via surface-initiated atom transfer radical polymerizations. Macromolecules 2005, 38 (18), 7867-7871.
  2. Zubair, M.; Jose, J.; Al-Harthi, M. A. Evaluation of mechanical and thermal properties of microwave irradiated poly (styrene-co-methyl methacrylate)/graphene nanocomposites. Compos. Interfaces 2015, 22 (7), 595-610.