Stearic acid hydrazide, 97%, Thermo Scientific Chemicals
Stearic acid hydrazide, 97%, Thermo Scientific Chemicals
Stearic acid hydrazide, 97%, Thermo Scientific Chemicals
Stearic acid hydrazide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Stearic acid hydrazide, 97%, Thermo Scientific Chemicals
CAS: 4130-54-5 | C18H38N2O | 298.515 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18291.30250 g
Catalog number ALFA18291.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (Aqueous acid-base Titration)≥47.5% to ≤49.5%