N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Acetylcaprolactam, 99%, Thermo Scientific Chemicals
CAS: 1888-91-1 | C8H13NO2 | 155.20 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18257.22100 g
Catalog number ALFA18257.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS336624-09-0
IUPAC Nametetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular FormulaC22H20N2Na4O10
InChI KeyZWSMLJACYSGFKD-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=C(OCCOC2=C(C=CC=C2)N(CC([O-])=O)CC([O-])=O)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to beige
Infrared spectrumConforms
Titration Complexometric>=94.0 %
Water=<12 % (K.F.)