3-Chloro-2-methylaniline, 98+%, Thermo Scientific Chemicals
3-Chloro-2-methylaniline, 98+%, Thermo Scientific Chemicals
3-Chloro-2-methylaniline, 98+%, Thermo Scientific Chemicals
3-Chloro-2-methylaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chloro-2-methylaniline, 98+%, Thermo Scientific Chemicals
CAS: 87-60-5 | C7H8ClN | 141.60 g/mol
Have Questions?
Catalog NumberQuantity
ALFA18221.30250 g
Catalog number ALFA18221.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material3-Chloro-2-methylaniline
CAS87-60-5
Health Hazard 1H302-H311-H315-H318-H335-H373
Health Hazard 2GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P361-P363-P501c