1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dibromotetrafluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 344-03-6 | C6Br2F4 | 307.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFA18135.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS13455-21-5
IUPAC Namepotassium dihydrate fluoride
Molecular FormulaFH4KO2
InChI KeyMZFXHSQFQLKKFD-UHFFFAOYSA-M
SMILESO.O.[F-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks
Assay19.65 to 20.70 % (F)
Appearance (Color)Colorless to white
It is used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a chemical intermediate.
Solubility
Solubility in methanol is almost transparent.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. A. Keith. Jameson.; Cynthia J. Jameson.Absolute temperature dependence of chemical shifts of lock solvents. Tetramethylsilane, hexafluorobenzene, and 1,4-dibromotetrafluorobenzene. Am. Chem. Soc.1973,95(26), 8559-8561.
  2. Alessandra Forni. Experimental and Theoretical Study of the Br···N Halogen Bond in Complexes of 1,4-Dibromotetrafluorobenzene with Dipyridyl Derivatives.J. Phys. Chem. A,2009,113(14), 3403-3412.