Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Isopropyl tetradecanoate, 98%, Thermo Scientific Chemicals
CAS: 110-27-0 | C17H34O2 | 270.457 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
2500 mL
Catalog number ALFA18111.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropyl myristate
CAS110-27-0
Recommended StorageAmbient temperatures
Density0.85
EINECS Number203-751-4
View more
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isopropyl tetradecanoate is used as a solvent in perfume products and the oil phase in the preparation of formulations. It finds application in eye makeup, nail care products, soaps and detergents, hair care products, shaving products and other skin care products. It behaves as a non-aqueous component of the two-phase mouthwash, Dentyl pH, thereby it removes bacteria from the oral cavity.
Solubility
Miscible with alcohol. Immiscible with water and glycerol.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Bashah, N. A. A.; Othman, M. R.; Aziz, N. Feed Forward Neural Network Model for Isopropyl Myristate Production in Industrial-scale Semi-batch Reactive Distillation Columns. Int. J. Eng. Sci. 2015, 11, 59-65.
  2. Vadgama, R. N.; Odaneth, A. A.; Lali, A. M. Green synthesis of isopropyl myristate in novel single phase medium Part II: Packed bed reactor (PBR) studies. Biotechnol. Rep. 2015, 8, 105-109.