2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H7Br2N | 204.893 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1000 g
Catalog number ALFA17625.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS118-93-4
IUPAC Name1-(2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H8O2
InChI KeyJECYUBVRTQDVAT-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.5570-1.5610 @ 20?C
Appearance (Color)Clear colorless to pale yellow to yellow to orange
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. O'Connor, L. J.; Cazares-Körner, C.; Saha, J.; Evans, C. N.; Stratford, M. R.; Hammond, E. M.; Conway, S. J. Efficient synthesis of 2-nitroimidazole derivatives and the bioreductive clinical candidate Evofosfamide (TH-302). Org. Chem. Front. 2015, 2 (9), 1026-1029.
  2. Maity, N.; Mandal, A.; Nandi, A. K. Interface engineering of ionic liquid integrated graphene in poly(vinylidene fluoride) matrix yielding magnificent improvement in mechanical, electrical and dielectric properties. Polymer 2015, 65, 154-167.