Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(4-hydroxyphenyl) sulfone, 99%, Thermo Scientific Chemicals
CAS: 80-09-1 | C12H10O4S | 250.27 g/mol
Have Questions?
Catalog NumberQuantity
ALFA17342.30250 g
Catalog number ALFA17342.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS480-40-0
IUPAC Name5,7-dihydroxy-2-phenyl-4H-chromen-4-one
Molecular FormulaC15H10O4
InChI KeyRTIXKCRFFJGDFG-UHFFFAOYSA-N
SMILESOC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point284°C to 290°C
Infrared spectrumConforms
Loss on drying=<1.0 %
Appearance (Color)Yellow
HPLC>=99.0 %
View more
It is a reactant used in epoxy reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a reactant used in epoxy reactions.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Takashi Ikegami.; Takashi Mukawa.; Hiroyuki Nariai.; Toshifumi Takeuchia. Bisphenol A-recognition polymers prepared by covalent molecular imprinting. Anal. Chim. Acta. 2004, 504 (1),131-135.
  2. Hiroshi Ishibashi.; Naoko Watanabe.; Naomi Matsumura.; Masashi Hirano.; Yukiko Nagao.; Hideki Shiratsuchi.; Shinya Kohra.; Shin-ichi Yoshihara.; Koji Arizonoa. Toxicity to early life stages and an estrogenic effect of a bisphenol A metabolite, 4-methyl-2,4-bis(4-hydroxyphenyl)pent-1-ene on the medaka (Oryzias latipes). Life Sci.. 2005, 77 (21),2643-2655.