o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals
o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals
o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals
o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
o-Dianisidine dihydrochloride, 98%, Thermo Scientific Chemicals
CAS: 20325-40-0 | C14H18Cl2N2O2 | 317.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALFA17175.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Materialo-Dianisidine dihydrochloride
CAS20325-40-0
Health Hazard 1H302-H314-H335-H350
Health Hazard 2GHS H Statement
H350-H302
Health Hazard 3P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3'-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is suitable for use in ELISA procedures. In neurohistochemistry o-Dianisidine dihydrochloride has demonstrated perikaryal labeling as well as retrograde and anterograde transport to the primary olfactory cortex. It has also been used for a paper based-micro fluidic device to semi-quantitatively measure lactate, uric acid, and glucose. 3,3′-Dimethoxybenzidine is used principally as an intermediate in the production of commercial bisazobiphenyl dyes for coloring textiles, paper, plastic, rubber, and leather.
Solubility
Soluble in water (50 mg/ml). It dissolves in ethyl alcohol ,acid and in organic solvent.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
RUO – Research Use Only
General References:
  1. Yanfen Peng.; Zhiqiang Gao.Amplified detection of microRNA based on ruthenium oxide nanoparticle-initiated deposition of an insulating film.Anal Chem.2011,83(3), 820-827.
  2. Wei Shen.; Huimin Deng.; Yuqian Ren, Zhiqiang Gao. A label-free microRNA biosensor based on DNAzyme-catalyzed and microRNA-guided formation of a thin insulating polymer film.Biosens Bioelectron.2013,44,171-176 .