3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
CAS: 609-99-4 | C7H4N2O7 | 228.116 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
50 g
250 g
Catalog number ALFA16988.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS4044-65-9
IUPAC Name1,4-diisothiocyanatobenzene
Molecular FormulaC8H4N2S2
InChI KeyOMWQUXGVXQELIX-UHFFFAOYSA-N
SMILESS=C=NC1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to yellow to beige to light grey
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Melting point129°C to 133°C
GC>=98.5 %
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.
Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
RUO – Research Use Only
General References:
  1. Taipa, M. A.; Azevedo, A. M.; Grilo, A. L.; Couto, P. T.; Ferreira, F. A. G.; Fortuna, A. R. M.; Pinto, I. F.; Santos, R. M.; Santos, S. B. Student Collaboration in a Series of Integrated Experiments To Study Enzyme Reactor Modeling with Immobilized Cell-Based Invertase. J. Chem. Educ. 2015, 92 (7), 1238-1243.
  2. Garcia-Moscoso, J. L.; Teymouri, A.; Kumar, S. Kinetics of Peptides and Arginine Production from Microalgae (Scenedesmus sp.) by Flash Hydrolysis. Ind. Eng. Chem. Res. 2015, 54 (7), 2048-2058.