Boric acid, 98%, Thermo Scientific Chemicals
Boric acid, 98%, Thermo Scientific Chemicals
Boric acid, 98%, Thermo Scientific Chemicals
Boric acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, 98%, Thermo Scientific Chemicals
CAS: 10043-35-3 | BH3O3 | 61.83 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16624.0B1000 g
Catalog number ALFA16624.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Chemical Identifiers
CAS2495-39-8
IUPAC Namesodium prop-2-ene-1-sulfonate
Molecular FormulaC3H5NaO3S
InChI KeyDIKJULDDNQFCJG-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration after Ion exchange>=94 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Boric Acid is a commonly used reagent in molecular biology for the preparation of buffers such as Tris Borate EDTA and has applications in capillary and gel electrophoresis. This compound has also been employed in a multitude of applictions, from weatherproofing fabrics to hardening steel. A commonly used molecular biology reagent that exhibits some antifungal activity
Solubility
Soluble in glycerol, ether, alcohol, methanol, liquid ammonia, acetone (slightly), water (6.35 g/100 cc) at 30°C, water (27.6 g/100 cc) at 100°C, water (50 g/L) at 21°C, and water (1 g / 18 ml cold water).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Chen FT1,; Sternberg JC.Characterization of proteins by capillary electrophoresis in fused-silica columns: review on serum protein analysis and application to immunoassays.. Electrophoresis.. 1994, 15(1),13-21
  2. Tůma P1,; Málková K,; Wedellová Z,; Samcová E,; Stulík K.The use of capillary electrophoresis with contactless conductivity detection for monitoring of glycerol in adipose tissues during a sporting performance.. Electrophoresis. 2010, 31(10),2037-2043.