2-Methoxyphenol, 98+%, Thermo Scientific Chemicals
2-Methoxyphenol, 98+%, Thermo Scientific Chemicals
2-Methoxyphenol, 98+%, Thermo Scientific Chemicals
2-Methoxyphenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methoxyphenol, 98+%, Thermo Scientific Chemicals
2-Methoxyphenol, >-98%, C7H8O2, CAS Number-90-05-1, methylcatechol, guaiastil, 2-hydroxyanisole, 1-hydroxy-2-methoxybenzene, pyroguaiac acid, o-methoxyphenol, pyrocatechol monomethyl ether, phenol, 2-methoxy, guaicol, guaiacol, 1000g, >98%, 508112, 204 deg.C to 206 deg.C, CHEBI:28591, 1.129
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number ALFA16319.30
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS142-03-0
IUPAC Name(acetyloxy)(hydroxy)alumanyl acetate
Molecular FormulaC4H7AlO5
InChI KeyHQQUTGFAWJNQIP-UHFFFAOYSA-K
SMILESCC(=O)O[Al](O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentAl2O3 content ≥ 28.0% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.
Solubility
Miscible with carbon tetrachloride, alcohol, chloroform, ether, oils, glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only
General References:
  1. Selvaraj, M.; Selvamani, A.; Alagarasi, A.; Gurulakshmi, M.; Krishnan, P. S.; Suthagar, K.; Shanthi, K. Hydrodeoxygenation of 2-Methoxyphenol Using NiO and NiO-MoO3/Mesoporous Aluminosilicate Catalysts: Effect of MoO3 and aluminum on Activity and Selectivity. Sci. Adv. Mater. 2015, 7 (4), 714-728.
  2. Leiva, K.; Sepulveda, C.; García, R.; Laurenti, D.; Vrinat, M.; Geantet, C.; Escalona, N. Kinetic study of the conversion of 2-methoxyphenol over supported Re catalysts: Sulfide and oxide state. Appl. Catal., A 2015, 505, 302-308.