1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrochloride monohydrate, 99%, Thermo Scientific Chemicals
CAS: 18851-33-7 | C12H11ClN2O | 234.683 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16254.1425 g
Catalog number ALFA16254.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS52449-43-1
IUPAC Namemethyl 2-(4-chlorophenyl)acetate
Molecular FormulaC9H9ClO2
InChI KeyWWIYGBWRUXQDND-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5220 to 1.5240 (20°C, 589 nm)
GC>=98.5 %
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,10-Phenanthroline monohydrochloride monohydrate is used as an analytical reagent for colorimetric determination of iron.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a tightly closed container in a cool, well-ventilated area away from air and moisture.
RUO – Research Use Only
General References:
  1. Liu, Y. L.; Xu, X. Y.; Sun, P. C.; Chen, T. H. N-doped porous carbon nanosheets with embedded iron carbide nanoparticles for oxygen reduction reaction in acidic media. Int. J. Hydrogen Energy 2015, 40 (13), 4531-4539.
  2. Vyprachticky, D.; Kankova, D.; Pokorna, V.; Kminek, I.; Dzhabarov, V.; Cimrova, V. Novel and Simple Synthesis of Brominated 1,10-Phenanthrolines. Aust. J. Chem. 2014, 67 (6), 915-921.