Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals
Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals
Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals
Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone, Thermo Scientific Chemicals
CAS: 108-05-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Catalog NumberQuantity
ALFA16247.AE100 mL
Catalog number ALFA16247.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS334-50-9
IUPAC Nametrihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
Molecular FormulaC7H22Cl3N3
InChI KeyLCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point260.0-265.0°C
Assay from Supplier's CofA≥99.0%
FormCrystals or crystalline powder
Appearance (Color)White
Water Content≤1.0%
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.
Solubility
Slightly miscible with water.
Notes
Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.
RUO – Research Use Only
General References:
  1. For example of mercuric acetate-catalyzed transesterification to long-chain vinyl esters, see: Org. Synth. Coll., 4, 977 (1963).
  2. For thermal [2+2] cycloaddition with chlorotrifluoroethylene, see: Org. Synth. Coll., 5, 235 (1973); and with chlorosulfonyl isocyanate to give a ß-lactam: Org. Synth. Coll., 8, 3 (1993).
  3. Dong, X.; Guan, X.; Jiang, Y.; Ma, J.; Zhang, M. Extension of the TraPPE-UA force field to the simulation of vapor-liquid phase equilibria of vinyl acetate system. J. Mol. Liq. 2015, 209, 520-525.
  4. Belbekhouche, S.; Picton, L.; Le Cerf, D.; Hamaide, T. Pullulan-Based Polymer Surfactants for Vinyl Acetate Miniemulsion Polymerization: Kinetics and Colloidal Stability Investigations. Macromol. Chem. Phys. 2015, 216 (18), 1879-1887.