Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals
Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals
Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals
Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
250 mL
Catalog number ALFA16084.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTri-n-butyl Phosphate
CAS126-73-8
Health Hazard 1H302-H315-H319-H351
Health Hazard 2GHS H Statement
H351-H302-H315
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Health Hazard 3P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
View more
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.
Solubility
Miscible with diethyl ether, benzene, carbon disulfide and ethanol. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases and alkali.
RUO – Research Use Only
General References:
  1. Vo, Q. N.; Hawkins, C. A.; Dang, L. X.; Nilsson, M.; Nguyen, H. D. Computational Study of Molecular Structure and Self-Association of Tri-n-butyl Phosphates in n-Dodecane. J. Phys. Chem. B 2015, 119 (4), 1588-1597.
  2. Rama, R.; Rout, A.; Venkatesan, K. A.; Antony, M. P.; Rao, P. R. V. Electrochemical behavior of Eu(III) in imidazolium ionic liquid containing tri-n-butyl phosphate and N, N-dihexyloctanamide ligands. J. Electroanal. Chem. 2015, 757, 36-43.