Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Tetrazolium Violet, 98+%
Thermo Scientific Chemicals
Tetrazolium Violet, 98+%
CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFA16008.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS29094-61-9
IUPAC NameN-[2-(4-{[(cyclohexylcarbamoyl)amino]sulfonyl}phenyl)ethyl]-5-methylpyrazine-2-carboxamide
Molecular FormulaC21H27N5O4S
InChI KeyZJJXGWJIGJFDTL-UHFFFAOYSA-N
SMILESCC1=CN=C(C=N1)C(=O)NCCC1=CC=C(C=C1)S(=O)(=O)NC(=O)NC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after ignition=<0.4 %
Appearance (Color)White to off-white
Loss on drying=<1 % (100°C, 3 hrs) (vacuum)
Heavy metals=<50 ppm