Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals
Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals
CAS: 206658-83-5 | C9H10N2NaO3+ | 217.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ALFA15775.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS26452-80-2
IUPAC Name2,4-dichloropyridine
Molecular FormulaC5H3Cl2N
InChI KeyTYPVHTOETJVYIV-UHFFFAOYSA-N
SMILESClC1=CC=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Form)Liquid
AppearanceMay darken on storage