n-Pentylbenzene, 96%, Thermo Scientific Chemicals
n-Pentylbenzene, 96%, Thermo Scientific Chemicals
n-Pentylbenzene, 96%, Thermo Scientific Chemicals
n-Pentylbenzene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Pentylbenzene, 96%, Thermo Scientific Chemicals
CAS: 538-68-1 | C11H16 | 148.249 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15734.065 g
Catalog number ALFA15734.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Materialn-Pentylbenzene
CAS538-68-1
Health Hazard 1H227
Health Hazard 2GHS H Statement
H227
Combustible liquid.
Health Hazard 3P210-P280-P370+P378q-P501c