4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
CAS: 100-09-4 | C8H8O3 | 152.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15697.30250 g
Catalog number ALFA15697.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS696-59-3
IUPAC Name2,5-dimethoxyoxolane
Molecular FormulaC6H12O3
InChI KeyGFISDBXSWQMOND-UHFFFAOYNA-N
SMILESCOC1CCC(OC)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5% (sum of isomers)
Refractive Index1.4160-1.4200 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
FormLiquid
4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzoic acid is used in oxidation and reduction of cytochrome c in solution through different self-assembled monolayers on gold electrodes using cyclic voltammetry. p-Anisic acid has antiseptic properties. It is also used as an intermediate in the preparation of more complex organic compounds.
Solubility
Soluble in water at 20°C 0.3g/L. Soluble in alcohol, ethyl acetate and ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Liu H, et al. Control of the electron transfer rate between cytochrome c and gold electrodes by the manipulation of the electrode's hydrogen bonding character.Langmuir.,2003,19(6), 2378-2387.
  2. Eggeling L and Sahm H. Degradation of coniferyl alcohol and other lignin-related aromatic compounds by Nocardia sp. DSM 1069.Arch. Microbiol.,1980,126(2), 141-148.