Methyl anthranilate, 99%, Thermo Scientific Chemicals
Methyl anthranilate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl anthranilate, 99%, Thermo Scientific Chemicals
CAS: 134-20-3 | C8H9NO2 | 151.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15681.30250 g
Catalog number ALFA15681.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1266615-79-5
IUPAC Name(4-methoxypyridin-3-yl)boronic acid
Molecular FormulaC6H8BNO3
InChI KeyYUTPAZKVEOJQCY-UHFFFAOYSA-N
SMILESCOC1=CC=NC=C1B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.