2-Chlorobenzylamine, 96%, Thermo Scientific Chemicals
2-Chlorobenzylamine, 96%, Thermo Scientific Chemicals
2-Chlorobenzylamine, 96%, Thermo Scientific Chemicals
2-Chlorobenzylamine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chlorobenzylamine, 96%, Thermo Scientific Chemicals
CAS: 89-97-4 | C7H8ClN | 141.598 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15594.1425 g
Catalog number ALFA15594.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS68-11-1
IUPAC Name2-sulfanylacetic acid
Molecular FormulaC2H4O2S
InChI KeyCWERGRDVMFNCDR-UHFFFAOYSA-N
SMILESOC(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Titration Iodimetric97.5 to 102.5 %
Refractive index1.5030 to 1.5050 (20°C, 589 nm)