Diiodomethane, 99%, stab., Thermo Scientific Chemicals
Diiodomethane, 99%, stab., Thermo Scientific Chemicals
Diiodomethane, 99%, stab., Thermo Scientific Chemicals
Diiodomethane, 99%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diiodomethane, 99%, stab., Thermo Scientific Chemicals
CAS: 75-11-6 | CH2I2 | 267.84 g/mol
Have Questions?
Catalog NumberQuantity
ALFA15457.1425 g
Catalog number ALFA15457.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration1.0N Sulfuric acid
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diiodomethane is used to determine the density of minerals and other solid samples due to its high density. It is used as an optical contact liquid to determine the refractive index of certain gemstones. In simmons-smith reaction, it acts as a reagent to generate methylene (carbine) free radical. It reacts with olefins to prepare cyclopropanes with high stereo specificity.
Solubility
Miscible with alcohol and ether. Slightly miscible with water.
Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with alkali metal salts, strong oxidizing agents, strong bases, metals, lithium, potassium, sodium, sodium oxides and alloys.
RUO – Research Use Only
General References:
  1. The combination with Zinc-copper couple, L09811, generates ICH2ZnI, the reagent for the classical Simmons-Smith cyclopropanation of alkenes: J. Am. Chem. Soc., 80, 5323 (1958); examples: Org. Synth. Coll., 5, 85 (1973). Review: Org. React., 20, 1 (1973). Alternative systems include Zn powder/ Cu(I) halide: J. Org. Chem., 35, 2057 (1970); ultrasound irradiation: Tetrahedron Lett., 23, 2729 (1982), thermal activation: Synth. Commun., 22, 2801 (1992), or catalytic TMS-Cl: J. Org. Chem., 59, 2671 (1994) with ordinary zinc; or Zn/Ag couple: Tetrahedron Lett., 4593 (1972); 3327 (1974). Other reagents include trialkylaluminums: J. Org. Chem., 50, 4412 (1985): Org. Synth. Coll., 8, 321 (1993) and the most widely-used modification, diethylzinc: Tetrahedron, 24, 53 (1968); J. Org. Chem., 42, 3031 (1977); for use in the ring-expansion of ketones, see: Org. Synth. Coll., 6, 327 (1988). For use of a boronic acid-derived chiral dioxaborolane ligand in enantioselective cyclopropanantion, see: Tetrahedron Lett., 37, 7925 (1996); Org. Synth., 76, 86 (1998):
  2. Selective ortho-methylation of phenols can be brought about by a one-pot method utilizing Et2Zn/ CH2I2 modification: Tetrahedron Lett., 30, 5215 (1989).
  3. Allylic bromides are converted to homoallylic iodides by the Simmons-Smith reagent in the presence of CuI.2LiI in THF: J. Org. Chem., 54, 5202 (1989).
  4. Wolcott, C. A.; Green, I. X.; Silbaugh, T. L.; Xu, Y.; Campbell, C. T. Energetics of Adsorbed CH2 and CH on Pt(111) by Calorimetry: The Dissociative Adsorption of Diiodomethane. J. Phys. Chem. C 2014, 118 (50), 29310-29321.
  5. Wang, Y.; Sang, D. K.; Du, Z.; Zhang, C.; Tian, M.; Mi, J. Interfacial Structures, Surface Tensions, and Contact Angles of Diiodomethane on Fluorinated Polymers. J. Phys. Chem. C 2014, 118 (19), 10143-10152.