2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
CAS: 93-25-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
Catalog number ALFA15211.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS92-51-3
IUPAC Name1,1'-bi(cyclohexane)
Molecular FormulaC12H22
InChI KeyWVIIMZNLDWSIRH-UHFFFAOYSA-N
SMILESC1CCC(CC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (U.K. specification)
Assay from Suppliers CofA≥97.5% (U.S. specification)
Identification (FTIR)Conforms (U.K. specification)
View more
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Hans-Adolf Arfmann; Wolf-Rainer Abraham. Microbial reduction of aromatic carboxylic acids. Zeitschrift fur Naturforschung C. 1993, 48,(1-2), 52-57.
  2. H.Pietilainena; T.Saesmaaab. HPLC determination of pindolol benzoate and pindolol 2-methoxyphenylacetate. Journal of Liquid Chromatography & Related Technologies. 1996, 19,(4), 583-591.