2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
CAS: 93-25-4 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
Catalog number ALFA15211.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS367-11-3
IUPAC Name1,2-difluorobenzene
Molecular FormulaC6H4F2
InChI KeyGOYDNIKZWGIXJT-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4420-1.4460 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.0%
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxyphenylacetic acid is used as pharmaceutical intermediate.
Solubility
Soluble in water. Also soluble in methanol, ether, ethanol and acetone.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Hans-Adolf Arfmann; Wolf-Rainer Abraham. Microbial reduction of aromatic carboxylic acids. Zeitschrift fur Naturforschung C. 1993, 48,(1-2), 52-57.
  2. H.Pietilainena; T.Saesmaaab. HPLC determination of pindolol benzoate and pindolol 2-methoxyphenylacetate. Journal of Liquid Chromatography & Related Technologies. 1996, 19,(4), 583-591.